Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Floor Mat Sticky mat, 24 in x 36 in, Blue, 30 sheets/pad, 4 pads/case (120 sheets), 45 μm sheet thickness, 1/8 in pad thickness, water-based acrylic adhesive
SDP

Catalog No. NC2166869
Encompass_Preferred

Multi-layer adhesive sticky mat (24 in x 36 in) engineered for contamination control in critical environments. Disposable peel-away polyethylene sheets capture foot-borne and wheel-borne contaminants, and numbered tabs make it easy to track remaining layers while revealing a fresh adhesive surface for continued protection.

  • Dimensions: 24 in x 36 in.

  • Color: Blue.

  • Material: polyethylene film.

  • Layers per pad: 30 sheets.

  • Pads per case: 4 pads (120 sheets).

  • Pad thickness: 1/8 in.

  • Sheet thickness: 45 μm per sheet.

  • Adhesive type: water-based acrylic.

  • Adhesive pH: 7-8.

  • RoHS compliant; free from heavy metals and hazardous chemicals.

  • Adhesive backing secures to carpet, tile, wood, and concrete without leaving residue.

  • Economical solution for clean rooms, hospitals, laboratories, manufacturing, and construction sites.
  • This item is not returnable. View return policy
    Catalog No. NC2166869 Supplier American Floor Mat Supplier No. BLUE
    May include imposed supplier surcharges.
    Add to Cart
    Add to Cart

    missing translation for 'validMainframeMsg'

    Multi-layer adhesive sticky mat (24 in x 36 in) engineered for contamination control in critical environments. Disposable peel-away polyethylene sheets capture foot-borne and wheel-borne contaminants, and numbered tabs make it easy to track remaining layers while revealing a fresh adhesive surface for continued protection.

  • Dimensions: 24 in x 36 in.

  • Color: Blue.

  • Material: polyethylene film.

  • Layers per pad: 30 sheets.

  • Pads per case: 4 pads (120 sheets).

  • Pad thickness: 1/8 in.

  • Sheet thickness: 45 μm per sheet.

  • Adhesive type: water-based acrylic.

  • Adhesive pH: 7-8.

  • RoHS compliant; free from heavy metals and hazardous chemicals.

  • Adhesive backing secures to carpet, tile, wood, and concrete without leaving residue.

  • Economical solution for clean rooms, hospitals, laboratories, manufacturing, and construction sites.
  • This item is not returnable. View return policy
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.