Learn More
Ergomat Inc Sticky Mat Frame, Gray, 24" x 50"
SDP
Catalog No. NC1325897
Encompass_Preferred
Sticky Mats are used to clean the soles of shoes to reduce traffic borne contaminants before entering critical areas with controlled sanitation. Sold as grey frame only.
Catalog No. NC1325897 Supplier Ergomat Inc Supplier No. SMFRAMEG
Sticky Mats are used to clean the soles of shoes to reduce traffic borne contaminants before entering critical areas with controlled sanitation. Sold as grey frame only.