Learn More
Ergomat Inc Sticky Mat, 300 Sheets, Blue
SDP
Catalog No. NC1325896
Encompass_Preferred
Sticky Mats are used to clean the soles of shoes to reduce traffic borne contaminants before entering critical areas with controlled sanitation. Sold as pack of 300 sheets. Pad size 46cm x 114cm (18” x 45”). 30 sheets per pad. Each sheet numbered to show remaining sheets.
Catalog No. NC1325896 Supplier Ergomat Inc Supplier No. SM1504BLUE
Sticky Mats are used to clean the soles of shoes to reduce traffic borne contaminants before entering critical areas with controlled sanitation. Sold as pack of 300 sheets. Pad size 46cm x 114cm (18” x 45”). 30 sheets per pad. Each sheet numbered to show remaining sheets.