Learn More
Ibidi USA 18 well bottomless µ-Slide, 15 per box
SDP
Catalog No. NC2119672
Encompass
An 18 well bottomless µ-Slide with a self-adhesive underside, allowing chosen substrates to be mounted. This design permits the mounting of a variety of bottom materials while providing an open well format for cell culture applications on any custom surfaces. Pcs./Box: 15 (individually packed)
This item is not returnable. View return policy
Catalog No. NC2119672 Supplier Ibidi USA Supplier No. 81818
An 18 well bottomless µ-Slide with a self-adhesive underside, allowing chosen substrates to be mounted. This design permits the mounting of a variety of bottom materials while providing an open well format for cell culture applications on any custom surfaces. Pcs./Box: 15 (individually packed)