Learn More
Ibidi USA sticky-Slide 8 Well, 15 pieces per box
SDP
Catalog No. 50305796
Encompass
The sticky Slide family allows you to perform cell culture experiments with custom¿ specific bottom materials like plastic sheets, glass coverslips, etc. The self-adhesive underside of the bottomless blank slide is easily adapted to your specific bottom substrate. The sticky¿Slide 8 Well provides a common open well format which is best suited for maximum sample access in a wide variety of experimental applications.Pcs./Box: 15 (individually packed)
This item is not returnable. View return policy
Catalog No. 50-305-796 Supplier Ibidi USA Supplier No. 80828
The sticky Slide family allows you to perform cell culture experiments with custom¿ specific bottom materials like plastic sheets, glass coverslips, etc. The self-adhesive underside of the bottomless blank slide is easily adapted to your specific bottom substrate. The sticky¿Slide 8 Well provides a common open well format which is best suited for maximum sample access in a wide variety of experimental applications.Pcs./Box: 15 (individually packed)