Learn More
Ibidi USA STICKY SLIDE 8 WELL HI 15 per box (individually packaged)
SDP
Catalog No. NC2029872
Encompass
A bottomless 8 well µ-Slide with high walls and a self-adhesive underside to mount a variety of bottom materials. Permits the mounting of a variety of bottom materials. With extra high individual walls to keep cross contamination between wells as low as possible while providing an open well format for cell culture applications on any custom surface. Pcs./Box: 15 (individually packed)
This item is not returnable. View return policy
Catalog No. NC2029872 Supplier Ibidi USA Supplier No. 80808
Add to Cart
Add to Cart
A bottomless 8 well µ-Slide with high walls and a self-adhesive underside to mount a variety of bottom materials. Permits the mounting of a variety of bottom materials. With extra high individual walls to keep cross contamination between wells as low as possible while providing an open well format for cell culture applications on any custom surface. Pcs./Box: 15 (individually packed)