Learn More
Ibidi USA sticky-Slide Chemotaxis
SDP
Catalog No. 50305777
Encompass
The sticky–Slide family allows you to perform cell culture experiments with custom– specific bottom materials like plastic sheets, glass coverslips, etc. The self-adhesive underside of the bottomless blank slide is easily adapted to your specific bottom substrate. The sticky–Slide Chemotaxis is a tool for investigation of chemotaxis and migration of non–adherent or adherent cells in gel matrices. The chamber’s geometry is optimized for analyzing chemotaxis by video microscopy. The linear concentration profile which is required for chemotactical movement is generated by diffusion and stable for at least 48 hours. The sticky version can be used to insert material into the large reservoirs or for assembling with custom–specific bottom materials, like glass coverslips or structured substrates. Sterile, 10 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-777 Supplier Ibidi USA Supplier No. 80328
The sticky–Slide family allows you to perform cell culture experiments with custom– specific bottom materials like plastic sheets, glass coverslips, etc. The self-adhesive underside of the bottomless blank slide is easily adapted to your specific bottom substrate. The sticky–Slide Chemotaxis is a tool for investigation of chemotaxis and migration of non–adherent or adherent cells in gel matrices. The chamber’s geometry is optimized for analyzing chemotaxis by video microscopy. The linear concentration profile which is required for chemotactical movement is generated by diffusion and stable for at least 48 hours. The sticky version can be used to insert material into the large reservoirs or for assembling with custom–specific bottom materials, like glass coverslips or structured substrates. Sterile, 10 pcs./box