Learn More
Ibidi USA sticky-Slide I 0.1 Luer, 15 pieces/box
SDP
Catalog No. 50305801
Encompass
The sticky Slide family allows you to perform cell culture experiments with custom¿ specific bottom materials like plastic sheets, glass coverslips, etc. The self-adhesive underside of the bottomless blank slide is easily adapted to your specific bottom substrate. The sticky¿Slide I Luer is designed for perfusion applications and applying defined shear stress and shear rates on cells inside the channel. 100 ¿m high channel, sterile, 15 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-801 Supplier Ibidi USA Supplier No. 81128
The sticky Slide family allows you to perform cell culture experiments with custom¿ specific bottom materials like plastic sheets, glass coverslips, etc. The self-adhesive underside of the bottomless blank slide is easily adapted to your specific bottom substrate. The sticky¿Slide I Luer is designed for perfusion applications and applying defined shear stress and shear rates on cells inside the channel. 100 ¿m high channel, sterile, 15 pcs./box