Learn More
Ibidi USA sticky-Slide I 0.6 Luer
SDP
Catalog No. 50305757
Encompass
The sticky–Slide family allows you to perform cell culture experiments with custom– specific bottom materials like plastic sheets, glass coverslips, etc. The self-adhesive underside of the bottomless blank slide is easily adapted to your specific bottom substrate. The sticky–Slide I Luer is designed for perfusion applications and applying defined shear stress and shear rates on cells inside the channel. 600 µm high channel, sterile, 15 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-757 Supplier Ibidi USA Supplier No. 80188
The sticky–Slide family allows you to perform cell culture experiments with custom– specific bottom materials like plastic sheets, glass coverslips, etc. The self-adhesive underside of the bottomless blank slide is easily adapted to your specific bottom substrate. The sticky–Slide I Luer is designed for perfusion applications and applying defined shear stress and shear rates on cells inside the channel. 600 µm high channel, sterile, 15 pcs./box