Learn More
Ibidi USA sticky-Slide VI 0.4
SDP
Catalog No. 50305785
Encompass
The sticky–Slide family allows you to perform cell culture experiments with custom– specific bottom materials like plastic sheets, glass coverslips, etc. The self-adhesive underside of the bottomless blank slide is easily adapted to your specific bottom substrate. The convenient six channel format is ideal for static cell cultivation and the application of standard immunofluorescence protocols for e.g. treatment, staining, and microscopy of living or fixed cells. The sticky–Slide VI 0.4 can also be connected to a pump and enables you to observe cells under flow conditions. 6 x 400 µm high channels, sterile, 15 pcs./box
This item is not returnable. View return policy
Catalog No. 50-305-785 Supplier Ibidi USA Supplier No. 80608
The sticky–Slide family allows you to perform cell culture experiments with custom– specific bottom materials like plastic sheets, glass coverslips, etc. The self-adhesive underside of the bottomless blank slide is easily adapted to your specific bottom substrate. The convenient six channel format is ideal for static cell cultivation and the application of standard immunofluorescence protocols for e.g. treatment, staining, and microscopy of living or fixed cells. The sticky–Slide VI 0.4 can also be connected to a pump and enables you to observe cells under flow conditions. 6 x 400 µm high channels, sterile, 15 pcs./box