Learn More
Cayman Chemical Stictic Acid, Purity: ≥95%, CAS: 549-06-4, 2.5 mg
SDP
Catalog No. 502933751
Encompass_Preferred
A depsidone that preferentially inhibits growth of HT-29 human colon adenocarcinoma cells over non-malignant cells (IC50s = 29.29 and 2,478.4 µg/ml, respectively)
Catalog No. 50-293-3751 Supplier Cayman Chemical Supplier No. 250172.5 MG
A depsidone that preferentially inhibits growth of HT-29 human colon adenocarcinoma cells over non-malignant cells (IC50s = 29.29 and 2,478.4 µg/ml, respectively)