Learn More
MSC Vikan Pipe Cleaning Brush, Stiff, 2.5-inch, Polyester, Black, 1 ea
Catalog No. 502817625
Encompass
This versatile pipe cleaning brush is ideal for thoroughly cleaning pipes, tubes, and hard-to-reach spaces in appliances like fish-sorting machines and meat mincers. It is compatible with any Vikan Euro threaded handle for extended reach.
  • Designed for food manufacturing, retail, and service, prioritizing hygiene and safety.
  • Hard, thick bristles dislodge stubborn soils such as baked-on dough and biofilms.
  • Curved bristle tip reaches into bends and corners.
  • Effective for pipe, tube, and valve cleaning.
  • Black color supports drain cleaning and HACCP color-coded systems.
  • Durable polypropylene and polyester construction ensures long-lasting use.
  • Color-coded for hygienic zoning and 5S lean programs.
  • Certified to EU 10/2011 for food contact.
  • Dishwasher and autoclave safe.
  • REACH compliant.
Catalog No. 50-281-7625 Supplier MSC Supplier No. 06730204
This versatile pipe cleaning brush is ideal for thoroughly cleaning pipes, tubes, and hard-to-reach spaces in appliances like fish-sorting machines and meat mincers. It is compatible with any Vikan Euro threaded handle for extended reach.
  • Designed for food manufacturing, retail, and service, prioritizing hygiene and safety.
  • Hard, thick bristles dislodge stubborn soils such as baked-on dough and biofilms.
  • Curved bristle tip reaches into bends and corners.
  • Effective for pipe, tube, and valve cleaning.
  • Black color supports drain cleaning and HACCP color-coded systems.
  • Durable polypropylene and polyester construction ensures long-lasting use.
  • Color-coded for hygienic zoning and 5S lean programs.
  • Certified to EU 10/2011 for food contact.
  • Dishwasher and autoclave safe.
  • REACH compliant.