Learn More
MSC Milwaukee Pipeline Inspection Reel, 325 ft. Stiff, 34 mm Camera Head, 1 ea
Catalog No. 5000514871
Encompass
This stiff pipeline inspection reel is engineered for long-run inspections in 3 in. to 10 in. sewer lines, offering enhanced maneuverability and clarity. Designed as part of a modular system, it integrates with a control hub and features a compact, high-definition self-leveling camera head for detailed pipe assessment.
  • Features a 34 mm 1080p hd self-leveling camera head
  • Supports up to 4X digital zoom and optimized light output
  • Includes a protective camera head sleeve and centering skids
  • Designed for 3 in. to 10 in. sewer lines
  • Equipped with a telescoping handle and wide wheelbase for ease of transport
Catalog No. 50-005-14871 Supplier MSC Supplier No. 57159758
This stiff pipeline inspection reel is engineered for long-run inspections in 3 in. to 10 in. sewer lines, offering enhanced maneuverability and clarity. Designed as part of a modular system, it integrates with a control hub and features a compact, high-definition self-leveling camera head for detailed pipe assessment.
  • Features a 34 mm 1080p hd self-leveling camera head
  • Supports up to 4X digital zoom and optimized light output
  • Includes a protective camera head sleeve and centering skids
  • Designed for 3 in. to 10 in. sewer lines
  • Equipped with a telescoping handle and wide wheelbase for ease of transport