Learn More
Med Vet International Stifle Retractor, Wallace, J0240WS, 5in, Each
Catalog No. 5000584385
Encompass_Preferred
The examination of the meniscus and intraarticular structure during anterior cruciate repair is a real necessity. This visualization often is difficult even in the largest of dogs. The stifle retractor allows gentle retraction between the femur and tibia, exposing the entire stifle joint to a thorough exam without damaging the joint surface. Stainless steel.
Catalog No. 50-005-84385 Supplier Med Vet International Supplier No. JOR240WS
The examination of the meniscus and intraarticular structure during anterior cruciate repair is a real necessity. This visualization often is difficult even in the largest of dogs. The stifle retractor allows gentle retraction between the femur and tibia, exposing the entire stifle joint to a thorough exam without damaging the joint surface. Stainless steel.