Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Stigmast-5-en-3-ol, 3-(hydrogen sulfate), (3β)- (trimethylamine) | 95.0% | 521.37 g·mol⁻¹ | C32H59NO4S | 10 MG
SDP

Catalog No. 5000295905
Encompass_Preferred

Sitosterol sulfate (trimethylamine) is the trimethylamine salt form of sitosterol sulfate intended for biological and pharmacological research. It is supplied as a solid and is provided with analytical documentation to support quality assessment.

  • Purity: 95.0%.
  • Molecular formula: C32H59NO4S.
  • Molecular weight: 521.37 g·mol⁻¹.
  • Supplied as a solid suitable for small-scale in vitro studies.
  • Includes COA and SDS for quality and safety information.

Catalog No. 50-002-95905 Supplier Medchemexpress LLC Supplier No. HYN10478A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sitosterol sulfate (trimethylamine) is the trimethylamine salt form of sitosterol sulfate intended for biological and pharmacological research. It is supplied as a solid and is provided with analytical documentation to support quality assessment.

  • Purity: 95.0%.
  • Molecular formula: C32H59NO4S.
  • Molecular weight: 521.37 g·mol⁻¹.
  • Supplied as a solid suitable for small-scale in vitro studies.
  • Includes COA and SDS for quality and safety information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.