Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Stigmasta-3,5-diene | 4970-37-0 | MFCD29036809 | >98.0% | 396.69 | C29H48 | 5 MG
SDP

Catalog No. 5000285120
Encompass_Preferred

Stigmasta-3,5-diene is a sterol-derived biochemical reagent (CAS 4970-37-0) used as a reference or assay reagent in biochemical and analytical research. It is supplied in small research quantities; follow the product certificate of analysis for confirmed purity and storage conditions.

  • Sterol-derived biochemical reagent for assay development and analytical use.
  • CAS number 4970-37-0 for unambiguous identification.
  • Molecular formula C29H48 and molecular weight 396.69.
  • Available in small vial sizes such as 1 mg and 5 mg for research applications.
  • Follow the certificate of analysis for storage, handling, and purity information.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-85120 Supplier Medchemexpress LLC Supplier No. HYW5878165MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stigmasta-3,5-diene is a sterol-derived biochemical reagent (CAS 4970-37-0) used as a reference or assay reagent in biochemical and analytical research. It is supplied in small research quantities; follow the product certificate of analysis for confirmed purity and storage conditions.

  • Sterol-derived biochemical reagent for assay development and analytical use.
  • CAS number 4970-37-0 for unambiguous identification.
  • Molecular formula C29H48 and molecular weight 396.69.
  • Available in small vial sizes such as 1 mg and 5 mg for research applications.
  • Follow the certificate of analysis for storage, handling, and purity information.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.