Learn More
Medchemexpress LLC Stigmasta-5,22-dien-3-ol, (3β,22E)- | 83-48-7 | 98.5% | 412.69 | 1 G
SDP
Catalog No. 5000428755
Encompass_Preferred
Stigmasterol is an orally active, immunomodulatory agent with anti-inflammatory and neuroprotective effects. It can cross the blood-brain barrier. It activates AMPK, inhibiting NF-κB and NLRP3 signaling pathways. Stigmasterol reduces microglia-mediated neuroinflammation and alleviates cognitive impairment and Alzheimer's disease. It also regulates M1/M2 polarization of microglia through the TLR4/NF-κB pathway, reducing neuropathic pain.
  • Orally active immunomodulatory agent
  • Exhibits anti-inflammatory effects
  • Neuroprotective properties
  • Crosses the blood-brain barrier
  • Activates AMPK
  • Inhibits NF-κB and NLRP3 signaling pathways
  • Reduces microglia-mediated neuroinflammation
  • Alleviates cognitive impairment and Alzheimer's disease
  • Regulates M1/M2 polarization of microglia
  • Reduces neuropathic pain
Catalog No. 50-004-28755 Supplier Medchemexpress LLC Supplier No. HYN01311G
Stigmasterol is an orally active, immunomodulatory agent with anti-inflammatory and neuroprotective effects. It can cross the blood-brain barrier. It activates AMPK, inhibiting NF-κB and NLRP3 signaling pathways. Stigmasterol reduces microglia-mediated neuroinflammation and alleviates cognitive impairment and Alzheimer's disease. It also regulates M1/M2 polarization of microglia through the TLR4/NF-κB pathway, reducing neuropathic pain.
  • Orally active immunomodulatory agent
  • Exhibits anti-inflammatory effects
  • Neuroprotective properties
  • Crosses the blood-brain barrier
  • Activates AMPK
  • Inhibits NF-κB and NLRP3 signaling pathways
  • Reduces microglia-mediated neuroinflammation
  • Alleviates cognitive impairment and Alzheimer's disease
  • Regulates M1/M2 polarization of microglia
  • Reduces neuropathic pain