Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Stigmastan-3-ol, (3β)- | 19466-47-8 | 98.5% | C29H52O | 25 MG
SDP

Catalog No. 5000357136
Encompass_Preferred

Stigmastan-3-ol, (3β)- is a chemical compound identified by CAS Number 19466-47-8, with a molecular formula of C29H52O. It is supplied as a white to off-white solid with a purity of 98.52%.

  • Identified by CAS number 19466-47-8
  • Molecular formula C29H52O
  • High purity of 98.52% (HPLC)
  • Supplied as a white to off-white solid
  • Recommended storage at 4°C, protected from light
  • When in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month, protected from light.

Catalog No. 50-003-57136 Supplier Medchemexpress LLC Supplier No. 25MGHY144513
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stigmastan-3-ol, (3β)- is a chemical compound identified by CAS Number 19466-47-8, with a molecular formula of C29H52O. It is supplied as a white to off-white solid with a purity of 98.52%.

  • Identified by CAS number 19466-47-8
  • Molecular formula C29H52O
  • High purity of 98.52% (HPLC)
  • Supplied as a white to off-white solid
  • Recommended storage at 4°C, protected from light
  • When in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.