Learn More
Apexbio Technology LLC Stigmasterol 83-48-7 100mg
SDP
Catalog No. 502240343
Encompass_Preferred
Stigmasterol (CAS 83-48-7) is a phytosterol derived from Glycine max (L ) Merr It structurally resembles cholesterol and is known to modulate cellular membrane fluidity and function Stigmasterol serves as a precursor in the biosynthesis of steroid hormones and has been reported to influence enzymes involved in lipid metabolism Due to its ability to integrate into lipid bilayers it is frequently utilized in research investigating cholesterol metabolism membrane dynamics and the effects of phytosterols on health-related biochemical pathways The compound is soluble in chloroform benzene ethyl acetate pyridine ethanol and acetone
This item is not returnable. View return policy
Catalog No. 50-224-0343 Supplier Apexbio Technology LLC Supplier No. N1281100
Stigmasterol (CAS 83-48-7) is a phytosterol derived from Glycine max (L ) Merr It structurally resembles cholesterol and is known to modulate cellular membrane fluidity and function Stigmasterol serves as a precursor in the biosynthesis of steroid hormones and has been reported to influence enzymes involved in lipid metabolism Due to its ability to integrate into lipid bilayers it is frequently utilized in research investigating cholesterol metabolism membrane dynamics and the effects of phytosterols on health-related biochemical pathways The compound is soluble in chloroform benzene ethyl acetate pyridine ethanol and acetone