Learn More
Selleck Chemical LLC Stigmasterol S2361-50mg
SDP
Catalog No. 503112614
Encompass_Preferred
Stigmasterol is an unsaturated plant sterol that exhibits anti-cancer anti-pyretic anti-inflammatory and immune-modulating effects
Catalog No. 50-311-2614 Supplier Selleck Chemical LLC Supplier No. S236150MG
Stigmasterol is an unsaturated plant sterol that exhibits anti-cancer anti-pyretic anti-inflammatory and immune-modulating effects