Learn More
AdipoGen Stigmasterol (5 g)
SDP
Catalog No. 502054638
Encompass_Preferred
Chemical. CAS: 83-48-7. Formula: C29H48O. MW: 412.69. Stigmasterol is a phytosterol with chemical structure similar to cholesterol and anti-hypercholestrolemic activity. It exhibits anticancer, anti-inflammatory, antioxidant, antidiabetic, neuroprotective and immune-modulating properties. Stigmasterol exhibits antitumor effects in cancer cells mediated via inhibition of cell migration, cell cycle arrest, apoptosis/autophagy induction and angiogenesis inhibition. Stigmasterol is a potent in vitro antagonist of FXR (farnesoid X receptor), a DNA polymerase beta inhibitor and targets the GLUT4 glucose transporter. It has anti-osteoarthritic effects, by decreasing the expression of matrix metalloproteinases. Stigmasterol shows acetylcholinesterase inhibitory activity and also enriched glutamatergic synapses in cultures of brain neurons.
Catalog No. 50-205-4638 Supplier AdipoGen Supplier No. CDXS0267G005
Chemical. CAS: 83-48-7. Formula: C29H48O. MW: 412.69. Stigmasterol is a phytosterol with chemical structure similar to cholesterol and anti-hypercholestrolemic activity. It exhibits anticancer, anti-inflammatory, antioxidant, antidiabetic, neuroprotective and immune-modulating properties. Stigmasterol exhibits antitumor effects in cancer cells mediated via inhibition of cell migration, cell cycle arrest, apoptosis/autophagy induction and angiogenesis inhibition. Stigmasterol is a potent in vitro antagonist of FXR (farnesoid X receptor), a DNA polymerase beta inhibitor and targets the GLUT4 glucose transporter. It has anti-osteoarthritic effects, by decreasing the expression of matrix metalloproteinases. Stigmasterol shows acetylcholinesterase inhibitory activity and also enriched glutamatergic synapses in cultures of brain neurons.