Learn More
Medchemexpress LLC Stigmasterol glucoside | 19716-26-8 | MFCD20274842 | 574.8 | 1 MG
SDP
Catalog No. 5000414995
Encompass_Preferred
Stigmasterol glucoside is a sterol compound and a 5α-reductase inhibitor with an IC50 value of 27.2 μM. This compound has demonstrated various biological activities.
  • It has anti-inflammatory properties.
  • It possesses antioxidant effects.
  • It exhibits anti-tumor activities.
Catalog No. 50-004-14995 Supplier Medchemexpress LLC Supplier No. HYN12001MG
Stigmasterol glucoside is a sterol compound and a 5α-reductase inhibitor with an IC50 value of 27.2 μM. This compound has demonstrated various biological activities.
  • It has anti-inflammatory properties.
  • It possesses antioxidant effects.
  • It exhibits anti-tumor activities.