Learn More
Kyfora Bio STEMS (Stigmasterol Hemisuccinate) 1 GR
Catalog No. 502559812
Encompass
Stigmasterol hemisuccinate (STEMS) is a stigmasterol derivative that is responsive to changes in pH. Stigmasterol is a naturally occurring phytosterol lipid that is growing in the bioprocessing space for its ability to improve lipid nanoparticle (LNP)-based mRNA transfection efficiency. Conjugation to hemiscuccinate has been shown to improve the cellular uptake in acidic microenvironments, such as those around tumors.Key application(s): LNP, acid-delivery systems
Catalog No. 50-255-9812 Supplier Kyfora Bio Supplier No. 144021
Stigmasterol hemisuccinate (STEMS) is a stigmasterol derivative that is responsive to changes in pH. Stigmasterol is a naturally occurring phytosterol lipid that is growing in the bioprocessing space for its ability to improve lipid nanoparticle (LNP)-based mRNA transfection efficiency. Conjugation to hemiscuccinate has been shown to improve the cellular uptake in acidic microenvironments, such as those around tumors.Key application(s): LNP, acid-delivery systems