Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Stikit™ Paper Disc Roll

Catalog No. 19072014 Shop All 3M Products
Change view
Click to view available options
Material:
Aluminum Oxide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-072-014 Aluminum Oxide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-014 Supplier 3M™ Supplier No. 7000119551
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Stikit™ Paper Disc Roll offers a versatile disc with a convenient adhesive backing ready for quick and easy attachment to a 3M™ Stikit™ backup pad (sold separately).

  • Adhesive backing allows for quick application and easy removal
  • Convenient disc rolls eliminate the need to remove disc liners from individual discs
  • Strong, C-weight paper backing provides extra durability when compared to lighter weight paper backings
  • Roll offers quick access to a disc and eliminates disc liners.
  • Aluminum oxide mineral is durable, cuts fast, and performs well on many surfaces
  • Load-resistant coating helps prevent premature loading of the disc surface to maximize cut and extend the life of the abrasive grains
TRUSTED_SUSTAINABILITY

Specifications

Product Type Paper Disc Roll
For Use With (Application) Finishing, Grinding, Sanding
Color Gold
Material Aluminum Oxide
Quantity 100 Discs/Roll, 4/Rolls/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.