Learn More
MSC Vikan Hand Brush, 8 Inch Length, Stiff, White, 1 ea
Catalog No. 502866072
Encompass
This Vikan hand brush is designed for effective cleaning, featuring a stiff bristle material for challenging tasks.
  • Made with polyester bristles and a plastic block
  • Stiff bristle firmness for thorough scrubbing
  • White bristle color
  • Overall length of 8 in. with a 1.3 in. bristle length
  • FDA compliant for use in regulated environments
  • Manufactured in Denmark
Catalog No. 50-286-6072 Supplier MSC Supplier No. 69813251
This Vikan hand brush is designed for effective cleaning, featuring a stiff bristle material for challenging tasks.
  • Made with polyester bristles and a plastic block
  • Stiff bristle firmness for thorough scrubbing
  • White bristle color
  • Overall length of 8 in. with a 1.3 in. bristle length
  • FDA compliant for use in regulated environments
  • Manufactured in Denmark