Learn More
Chemglass Life Sciences Hickman Still Head, Minum-Ware
SDP
Catalog No. 50873329
Encompass_Preferred
Hickman Still Head, Minum-Ware. Unique design of the Hickman still makes it both the collection vessel and condenser for simple fractional distillations. The threaded side port permits easy removal of distillate via a syringe or Pasteur pipette. Side port has a 8-425 thread. Still has an upper threaded outer joint and lower inner joint, both 14/10. Supplied complete with open-top screw cap (MW-102-33) and Viton o-ring (MW-100-08) for 14/10 joint and PTFE faced septa (MW-104-01) and open-top screw cap (MW-102-11) for 8-425 sidearm.
Catalog No. 50-873-329 Supplier Chemglass Life Sciences Supplier No. MW5001
Hickman Still Head, Minum-Ware. Unique design of the Hickman still makes it both the collection vessel and condenser for simple fractional distillations. The threaded side port permits easy removal of distillate via a syringe or Pasteur pipette. Side port has a 8-425 thread. Still has an upper threaded outer joint and lower inner joint, both 14/10. Supplied complete with open-top screw cap (MW-102-33) and Viton o-ring (MW-100-08) for 14/10 joint and PTFE faced septa (MW-104-01) and open-top screw cap (MW-102-11) for 8-425 sidearm.