Learn More
VWR International Stilla® Ruby Chip Pack of 12
Supplier:  VWR International 77392266
Encompass
The Ruby Chip features cutting-edge, patented microfluidic technology that integrates the entire digital PCR workflow into a single fully automation-compatible consumable.Capacity: 16 samples/chipInput volume: 5 µl/chamberDroplets per sample: Up to 17000Dynamic range: ∼4.7 logFully compatible with integration to automated liquid handling equipment
Catalog No. NC2887569