Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Stille Pattern Rib Shears

Catalog No. 21124629 Shop All Surgical Design Products
Change view
Click to view available options
Length (English):
8.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
21-124-629 8.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-124-629 Supplier Surgical Design™ Supplier No. ATPM009
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stille pattern rib shears, 8.5 in., feature heavy-duty blades for clean, controlled cuts through ribs and cartilage during thoracic cavity opening.

  • 8.5-in. Stille pattern rib shears engineered specifically for cutting ribs, cartilage, and dense thoracic structures during autopsy
  • Heavy-duty curved blades provide strong cutting power while reducing operator fatigue
  • Compound-action mechanism enhances leverage for smooth cutting through thick rib plates
  • Offset blade angle improves visibility and access when opening the chest cavity
  • Constructed from premium surgical-grade stainless steel for durability under rigorous postmortem use
  • Serrated cutting edge minimizes slippage and ensures cleaner cuts through tough tissue and bone
  • Ergonomically designed handles for secure, comfortable grip, even with gloved or wet hands
  • Fully autoclavable, corrosion-resistant, and suitable for high-volume pathology or forensic labs
  • Ideal for thoracic cavity entry, organ removal protocols, and forensic examinations
  • Opening the rib cage during thoracic autopsy
  • Cutting through ribs, costal cartilage, and sternum-adjacent structures
  • Performing medicolegal postmortem examinations requiring rapid chest exposure
  • Suitable for pathology departments, medical examiners, veterinary necropsy, and teaching labs

Specifications

Product Type Bone-cutting Forceps
Blade Material Stainless Steel
Material Stainless Steel
Length (English) 8.5 in.
Length (Metric) 21.60 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.