Learn More
Sklar Instruments Stille Twist Drill Bit, Sklar, 2mm Diameter, Stainless Steel. EA/1.
SDP
Catalog No. 501438311 Shop All Sklar Instruments Products
Encompass_Preferred
Stille Twist Drill Bit, Sklar, 2mm Diameter, Stainless Steel. Sklar's Stille twist drill bit is is used in procedures that require creating or enlarging holes in bones for pins or wire. The drill is handheld and cranked manually. The drill bits sold seperately are available in a variety of sizes, for use in Stille drill product # 40-1486.
This item is not returnable. View return policy
Catalog No. 50-143-8311 Supplier Sklar Instruments Supplier No. 401498
Stille Twist Drill Bit, Sklar, 2mm Diameter, Stainless Steel. Sklar's Stille twist drill bit is is used in procedures that require creating or enlarging holes in bones for pins or wire. The drill is handheld and cranked manually. The drill bits sold seperately are available in a variety of sizes, for use in Stille drill product # 40-1486.