Learn More
Aviva Systems Biology Corp STIM1 Antibody - middle region (ARP45434_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45434_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STIM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human STIM1. Alternate names: D11S4896E, GOK, STIM1LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-53864