Learn More
Grass Technologies STIMULATING OR RECORDING DIGIT
Supplier:  Grass Technologies 6032TP
Encompass
stimulating or recording digital ring electrodes, stainless steel, 19" cable length and 1.5mm female touchproof connectors. 1 pair/pkg.
Catalog No. NC1960414