Learn More
Med Vet International Stimuplex A Needle, 21g x 4" (100mm), J1531C, Each
Catalog No. 5000583148
Encompass_Preferred
Stimuplex A • Non-ultrasound guided for nerve stimulation and injection • With depth markings to identify puncture depth • 30° facet bevel Stimuplex A Needle, 21g x 4" (100mm)
Catalog No. 50-005-83148 Supplier Med Vet International Supplier No. JOR1531C
Stimuplex A • Non-ultrasound guided for nerve stimulation and injection • With depth markings to identify puncture depth • 30° facet bevel Stimuplex A Needle, 21g x 4" (100mm)