Learn More
Med Vet International Stimuplex A Needle, 22g x 2" (50mm), J1531D, Each
Catalog No. 5000581634
Encompass_Preferred
Stimuplex A • Non-ultrasound guided for nerve stimulation and injection • With depth markings to identify puncture depth • 30° facet bevel Stimuplex A Needle, 22g x 2" (50mm)
Catalog No. 50-005-81634 Supplier Med Vet International Supplier No. JOR1531D
Stimuplex A • Non-ultrasound guided for nerve stimulation and injection • With depth markings to identify puncture depth • 30° facet bevel Stimuplex A Needle, 22g x 2" (50mm)