Learn More
TARGETMOL CHEMICALS INC STING ACTIVATOR COMPOUND 10MG
SDP
Catalog No. 502576189
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. STING activator Compound 53 based on an oxindole core structure demonstrated robust on-target functional activation of STING (human EC50 185 nM) in immortalised and primary cells and a cytokine induction fingerprint consistent with STING activation. purity: 98%
Catalog No. 50-257-6189 Supplier TARGETMOL CHEMICALS INC Supplier No. T985310MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. STING activator Compound 53 based on an oxindole core structure demonstrated robust on-target functional activation of STING (human EC50 185 nM) in immortalised and primary cells and a cytokine induction fingerprint consistent with STING activation. purity: 98%