Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC STING Agonist-12 | 2259624-71-8 | 98.3% | 490.88 | 50 MG
SDP

Catalog No. 5000362460
Encompass_Preferred

STING agonist-12 is a potent, orally active human STING activator with an EC50 of 185 nM. It demonstrates excellent pan-polymorph activity across STING proteins in HEK293T cells. This compound is well-absorbed in animal models, exhibiting a short terminal half-life. For research use only.

  • Potent, orally active human STING activator
  • EC50 of 185 nM against STING
  • Excellent pan-polymorph activity
  • Not active at mouse STING
  • Well-absorbed in animal models
  • Short terminal half-life
  • Appearance: Solid, light yellow to yellow
  • Shipping: Room temperature (continental US)
  • Storage: 4°C, protect from light (solid)
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (protect from light)

Catalog No. 50-003-62460 Supplier Medchemexpress LLC Supplier No. HY14701050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

STING agonist-12 is a potent, orally active human STING activator with an EC50 of 185 nM. It demonstrates excellent pan-polymorph activity across STING proteins in HEK293T cells. This compound is well-absorbed in animal models, exhibiting a short terminal half-life. For research use only.

  • Potent, orally active human STING activator
  • EC50 of 185 nM against STING
  • Excellent pan-polymorph activity
  • Not active at mouse STING
  • Well-absorbed in animal models
  • Short terminal half-life
  • Appearance: Solid, light yellow to yellow
  • Shipping: Room temperature (continental US)
  • Storage: 4°C, protect from light (solid)
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.