Learn More
Selleck Chemical LLC STING agonist-15mg
SDP
Catalog No. 502173453
Encompass_Preferred
STING agonist-1 (G10) is a novel human-specificSTING agonist that triggersIFN regulatory factor 3 (IRF3)/ type I interferon (IFN)-associated transcription in human fibroblasts. STING agonist-1 (G10) potently reduces growth of Chikungunya virus (CHIKV) with IC90 of 8.01 ?M and blocks replication of Alphavirus species Venezuelan Equine Encephalitis Virus (VEEV) with IC90 of 24.57 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3453 Supplier Selleck Chemical LLC Supplier No. S89545MG
Add to Cart
Add to Cart
STING agonist-1 (G10) is a novel human-specificSTING agonist that triggersIFN regulatory factor 3 (IRF3)/ type I interferon (IFN)-associated transcription in human fibroblasts. STING agonist-1 (G10) potently reduces growth of Chikungunya virus (CHIKV) with IC90 of 8.01 ?M and blocks replication of Alphavirus species Venezuelan Equine Encephalitis Virus (VEEV) with IC90 of 24.57 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.