Learn More
Medchemexpress LLC STING agonist-17 | 2816929-47-0 | 96.2% | C43H53N13O8 | 10 MM * 1 ML
SDP
Catalog No. 5000356151
Encompass_Preferred
STING agonist-17 is a potent STING agonist with an IC50 value of 0.062 nM, demonstrating anti-cancer activity for tumor immunization.
  • Potent STING agonist with an IC50 of 0.062 nM.
  • Exhibits anti-cancer activity through tumor immunization.
  • Inhibits several CYP isozymes, including CYP3A4 (IC50 = 4.2 μM).
  • Induces IFN-β secretion with an EC50 of 2.0 nM in THP-1 dual cells.
  • Activates key signaling factors such as STING, TBK1, IRF3, and STAT1.
  • Demonstrates an inhibitory effect on tumor growth in in vivo models.
Catalog No. 50-003-56151 Supplier Medchemexpress LLC Supplier No. HY14332010MM/1ML
STING agonist-17 is a potent STING agonist with an IC50 value of 0.062 nM, demonstrating anti-cancer activity for tumor immunization.
  • Potent STING agonist with an IC50 of 0.062 nM.
  • Exhibits anti-cancer activity through tumor immunization.
  • Inhibits several CYP isozymes, including CYP3A4 (IC50 = 4.2 μM).
  • Induces IFN-β secretion with an EC50 of 2.0 nM in THP-1 dual cells.
  • Activates key signaling factors such as STING, TBK1, IRF3, and STAT1.
  • Demonstrates an inhibitory effect on tumor growth in in vivo models.