Learn More
Medchemexpress LLC STING agonist-22 | 2408723-12-4 | C40H48N14O6 | 50 MG
SDP
Catalog No. 5000372318
Encompass_Preferred
STING agonist-22 (CF501) is a potent non-nucleotide STING agonist. It acts as an adjuvant by activating STING to induce type I interferon (IFN-I) response and proinflammatory cytokine production. This product boosts protein vaccines for potent, broad, and long-term immune protection, and is suitable for research on SARS-CoV-2 variants and sarbecovirus diseases.
  • Activates STING, inducing type I interferon (IFN-I) and proinflammatory cytokine production.
  • Boosts protein vaccines as an adjuvant.
  • Provides potent, broad, and long-term immune protection.
  • Suitable for SARS-CoV-2 variants and sarbecovirus research.
  • Rapidly activates innate immune responses in THP-1 cells.
  • Activates innate immunity in mice with acceptable safety.
  • Stimulates humoral and cellular immune responses.
Catalog No. 50-003-72318 Supplier Medchemexpress LLC Supplier No. HY15295550MG
STING agonist-22 (CF501) is a potent non-nucleotide STING agonist. It acts as an adjuvant by activating STING to induce type I interferon (IFN-I) response and proinflammatory cytokine production. This product boosts protein vaccines for potent, broad, and long-term immune protection, and is suitable for research on SARS-CoV-2 variants and sarbecovirus diseases.
  • Activates STING, inducing type I interferon (IFN-I) and proinflammatory cytokine production.
  • Boosts protein vaccines as an adjuvant.
  • Provides potent, broad, and long-term immune protection.
  • Suitable for SARS-CoV-2 variants and sarbecovirus research.
  • Rapidly activates innate immune responses in THP-1 cells.
  • Activates innate immunity in mice with acceptable safety.
  • Stimulates humoral and cellular immune responses.