Learn More
Medchemexpress LLC STING agonist-3 | 2138299-29-1 | >98.0% | 750.8 g/mol | C37H42N12O6 | 5 MG
SDP
Catalog No. 5000245211
Encompass_Preferred
diABZI STING Agonist 3 is a selective, non-nucleotide small-molecule agonist of the STING pathway used in preclinical research to activate innate immune signaling and study antitumor immunostimulatory effects. It is employed in biochemical and cell-based assays to probe STING activation and downstream interferon responses.
  • Activates STING signaling in biochemical and cell-based assays.
  • Demonstrates potent binding affinity and cellular activity.
  • Suitable for in vitro and preclinical in vivo studies.
  • Available in small research pack sizes for assay development.
  • High purity appropriate for research applications.
Catalog No. 50-002-45211 Supplier Medchemexpress LLC Supplier No. HY1036655MG
diABZI STING Agonist 3 is a selective, non-nucleotide small-molecule agonist of the STING pathway used in preclinical research to activate innate immune signaling and study antitumor immunostimulatory effects. It is employed in biochemical and cell-based assays to probe STING activation and downstream interferon responses.
  • Activates STING signaling in biochemical and cell-based assays.
  • Demonstrates potent binding affinity and cellular activity.
  • Suitable for in vitro and preclinical in vivo studies.
  • Available in small research pack sizes for assay development.
  • High purity appropriate for research applications.