Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC STING agonist-3 trihydrochloride | 98.4% | 860.19 | C37H45Cl3N12O6 | 10 MG
SDP

Catalog No. 5000244708
Encompass_Preferred

STING agonist-3 trihydrochloride is a selective, non-nucleotide small-molecule agonist of STING used for research in immunology and oncology. Supplied as the trihydrochloride salt in a solid, white to off-white form, it is characterized by molecular formula C37H45Cl3N12O6, molecular weight 860.19, and high purity (98.36%).

  • Selective non-nucleotide STING agonist with demonstrated activity in preclinical models.
  • Molecular formula C37H45Cl3N12O6; molecular weight 860.19.
  • High purity (98.36%) suitable for research applications.
  • Supplied as a solid; recommended dissolution in DMSO for in vitro use.
  • Storage recommendations: solid at 4°C (protect from light); in solution store at -80°C for long-term.

Catalog No. 50-002-44708 Supplier Medchemexpress LLC Supplier No. HY103665A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

STING agonist-3 trihydrochloride is a selective, non-nucleotide small-molecule agonist of STING used for research in immunology and oncology. Supplied as the trihydrochloride salt in a solid, white to off-white form, it is characterized by molecular formula C37H45Cl3N12O6, molecular weight 860.19, and high purity (98.36%).

  • Selective non-nucleotide STING agonist with demonstrated activity in preclinical models.
  • Molecular formula C37H45Cl3N12O6; molecular weight 860.19.
  • High purity (98.36%) suitable for research applications.
  • Supplied as a solid; recommended dissolution in DMSO for in vitro use.
  • Storage recommendations: solid at 4°C (protect from light); in solution store at -80°C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.