Learn More
Medchemexpress LLC STING agonist-3 trihydrochloride | 98.4% | 860.19 g/mol | C37H45Cl3N12O6 | 1 ML
SDP
Catalog No. 5000246864
Encompass_Preferred
STING agonist-3 trihydrochloride is a selective, non-nucleotide small-molecule stimulator of interferon genes (STING) supplied as a 10 mM solution in DMSO for research use. The compound is provided as the trihydrochloride salt with reported purity of 98.36% and molecular weight 860.19 g/mol; storage recommendations are solid at 4°C protected from light and under nitrogen, and in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Selective non-nucleotide STING agonist with pEC50 of 7.5.
  • Supplied as a 10 mM solution in DMSO, 1 mL vial.
  • Trihydrochloride salt form to aid handling and solubility.
  • High purity (98.36%) with supporting analytical data.
  • Intended for immuno-oncology and STING pathway research.
  • Documentation available: datasheet, COA, SDS, HNMR, LCMS.
Catalog No. 50-002-46864 Supplier Medchemexpress LLC Supplier No. HY103665A10MM 1ML
STING agonist-3 trihydrochloride is a selective, non-nucleotide small-molecule stimulator of interferon genes (STING) supplied as a 10 mM solution in DMSO for research use. The compound is provided as the trihydrochloride salt with reported purity of 98.36% and molecular weight 860.19 g/mol; storage recommendations are solid at 4°C protected from light and under nitrogen, and in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Selective non-nucleotide STING agonist with pEC50 of 7.5.
  • Supplied as a 10 mM solution in DMSO, 1 mL vial.
  • Trihydrochloride salt form to aid handling and solubility.
  • High purity (98.36%) with supporting analytical data.
  • Intended for immuno-oncology and STING pathway research.
  • Documentation available: datasheet, COA, SDS, HNMR, LCMS.