Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sting agonist-7 | 2767442-46-4 | 97.2% | C17H12N4O4 | 10MG
SDP

Catalog No. 5000258500
Encompass_Preferred

STING agonist-7 is a non-nucleotide agonist of the stimulator of interferon genes (STING) pathway used in research to probe STING-mediated immunology and inflammation. It binds selectively to mouse STING but not human STING and has limited cell membrane permeability. The compound is supplied at high purity and is DMSO-soluble for experimental applications.

  • Non-nucleotide STING agonist suitable for pathway studies.
  • Selective binding to mouse STING with minimal activity on human STING.
  • Poor cell membrane permeability, relevant for assay design.
  • High purity (~97.2%) for reproducible results.
  • Soluble in DMSO (62.5 mg/mL); ultrasonic aid recommended.
  • Storage: 4°C protected from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-58500 Supplier Medchemexpress LLC Supplier No. HY14389610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

STING agonist-7 is a non-nucleotide agonist of the stimulator of interferon genes (STING) pathway used in research to probe STING-mediated immunology and inflammation. It binds selectively to mouse STING but not human STING and has limited cell membrane permeability. The compound is supplied at high purity and is DMSO-soluble for experimental applications.

  • Non-nucleotide STING agonist suitable for pathway studies.
  • Selective binding to mouse STING with minimal activity on human STING.
  • Poor cell membrane permeability, relevant for assay design.
  • High purity (~97.2%) for reproducible results.
  • Soluble in DMSO (62.5 mg/mL); ultrasonic aid recommended.
  • Storage: 4°C protected from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.