Learn More
Medchemexpress LLC STING agonist-7 | 2767442-46-4 | 97.22% | C17H12N4O4 | 100 MG
SDP
Catalog No. 5000355289
Encompass_Preferred
STING agonist-7 is a non-nucleotide STING agonist that selectively binds to mouse STING, but not human STING. This compound poorly penetrates cell membranes. It is intended for research use only.
  • Selectively binds to mouse STING
  • Does not bind to human STING
  • Poorly penetrates cell membranes
  • Active in biochemical assays
  • Inactive in cellular reporter assays
Catalog No. 50-003-55289 Supplier Medchemexpress LLC Supplier No. HY143896100MG
STING agonist-7 is a non-nucleotide STING agonist that selectively binds to mouse STING, but not human STING. This compound poorly penetrates cell membranes. It is intended for research use only.
  • Selectively binds to mouse STING
  • Does not bind to human STING
  • Poorly penetrates cell membranes
  • Active in biochemical assays
  • Inactive in cellular reporter assays