Learn More
Medchemexpress LLC STING agonist-7 | 2767442-46-4 | 97.22% | C17H12N4O4 | 50 MG
SDP
Catalog No. 5000353303
Encompass_Preferred
STING agonist-7 is a non-nucleotide STING agonist. It selectively binds to mouse STING but not human STING and penetrates cell membranes poorly. In vitro, STING agonist-7 (compound 11) is active in biochemical assays but inactive in cellular reporter assays.
  • Non-nucleotide STING agonist
  • Selectively binds to mouse STING
  • Poor cell membrane penetration
  • Active in biochemical assays
  • Inactive in cellular reporter assays
  • Solid appearance
  • Color is light brown to brown
Catalog No. 50-003-53303 Supplier Medchemexpress LLC Supplier No. HY14389650MG
STING agonist-7 is a non-nucleotide STING agonist. It selectively binds to mouse STING but not human STING and penetrates cell membranes poorly. In vitro, STING agonist-7 (compound 11) is active in biochemical assays but inactive in cellular reporter assays.
  • Non-nucleotide STING agonist
  • Selectively binds to mouse STING
  • Poor cell membrane penetration
  • Active in biochemical assays
  • Inactive in cellular reporter assays
  • Solid appearance
  • Color is light brown to brown