Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC STING agonist-8 dihydrochloride | 2745723-90-2 | >98% | C41H48Cl2N14O4 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHY144168A

Encompass_Preferred

STING agonist-8 dihydrochloride, also known as compound 5-AB, is a potent Stimulator of Interferon Genes (STING) agonist with an EC50 of 27 nM in THP1-Dual KI-hSTING-R232 cells. This product is designated exclusively for research purposes, ideal for studying the STING pathway in innate immunity.

  • Potent STING agonist.
  • Supplied as a dihydrochloride salt, a white to off-white solid.
  • Soluble in DMSO at 5 mg/mL (5.74 mM) with sonication and heating.
  • Store at -20°C in a sealed container, protected from moisture.
  • Useful for immunology, inflammation, and drug discovery research.
  • Applied in in vitro and in vivo studies of STING activation.

Catalog No. 50-003-57276


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.