Learn More
Cell Signaling Technology STING (E9X7F) Rabbit Monoclonal Antibody, Human, Mouse, Unconjugated, 100 μl
SDP
Catalog No. 502730174
Encompass
The STING (E9X7F) Rabbit Monoclonal Antibody is a high-quality, recombinant primary antibody designed to recognize endogenous levels of total STING protein. Produced by immunizing animals with human STING recombinant protein, this monoclonal antibody reacts with an epitope near the carboxy terminus and is ideal for applications involving the study of the innate immune response to pathogenic cytoplasmic DNA.
  • Validated for immunofluorescence and flow cytometry applications.
  • Reacts with human and mouse species.
  • Provided in a stable formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Recommended storage at −20°C.
  • Exhibits sensitivity for total protein in both human and mouse cell lines, although some low-expressing mouse cell lines may show reduced sensitivity by immunofluorescence.
Catalog No. 50-273-0174 Supplier Cell Signaling Technology Supplier No. 90947S
The STING (E9X7F) Rabbit Monoclonal Antibody is a high-quality, recombinant primary antibody designed to recognize endogenous levels of total STING protein. Produced by immunizing animals with human STING recombinant protein, this monoclonal antibody reacts with an epitope near the carboxy terminus and is ideal for applications involving the study of the innate immune response to pathogenic cytoplasmic DNA.
  • Validated for immunofluorescence and flow cytometry applications.
  • Reacts with human and mouse species.
  • Provided in a stable formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Recommended storage at −20°C.
  • Exhibits sensitivity for total protein in both human and mouse cell lines, although some low-expressing mouse cell lines may show reduced sensitivity by immunofluorescence.