Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sting-In-3 | 2244881-69-2 | 99.8% | 316.35 | 100 MG
SDP

Catalog No. 5000349117
Encompass_Preferred

STING-IN-3 is an inhibitor of stimulator of interferon genes (STING). It efficiently inhibits both human STING (hsSTING) and mouse STING (mmSTING) by covalently targeting the predicted transmembrane cysteine residue 91. This action blocks the activation-induced palmitoylation of STING.

  • Biological activity: STING-IN-3 (0.02-2 μM) inhibits human STING and mouse STING, mediated IFNβ reporter activity in HEK293 cells.
  • Target: STING
  • Appearance: Solid
  • Color: Light yellow to green yellow

Catalog No. 50-003-49117 Supplier Medchemexpress LLC Supplier No. HY138683100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

STING-IN-3 is an inhibitor of stimulator of interferon genes (STING). It efficiently inhibits both human STING (hsSTING) and mouse STING (mmSTING) by covalently targeting the predicted transmembrane cysteine residue 91. This action blocks the activation-induced palmitoylation of STING.

  • Biological activity: STING-IN-3 (0.02-2 μM) inhibits human STING and mouse STING, mediated IFNβ reporter activity in HEK293 cells.
  • Target: STING
  • Appearance: Solid
  • Color: Light yellow to green yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.