Learn More
Medchemexpress LLC Sting-in-3 | 2244881-69-2 | 99.8% | 316.35 g·mol⁻¹ | C17H20N2O4 | 5 MG
SDP
Catalog No. 5000223310
Encompass_Preferred
STING-IN-3 is a small-molecule covalent inhibitor of the stimulator of interferon genes (STING) used for research applications. It inhibits both human and mouse STING by covalently targeting a predicted transmembrane cysteine residue, blocking STING activation and downstream signaling. Supplied as a powder or DMSO solution, it is intended for biochemical and cellular assay use.
Catalog No. 50-002-23310 Supplier Medchemexpress LLC Supplier No. HY1386835MG
STING-IN-3 is a small-molecule covalent inhibitor of the stimulator of interferon genes (STING) used for research applications. It inhibits both human and mouse STING by covalently targeting a predicted transmembrane cysteine residue, blocking STING activation and downstream signaling. Supplied as a powder or DMSO solution, it is intended for biochemical and cellular assay use.