Learn More
Medchemexpress LLC Sting-in-3 | 2244881-69-2 | 99.8% | C17H20N2O4 | 10MG
SDP
Catalog No. 5000259050
Encompass_Preferred
STING-IN-3 is a covalent small-molecule inhibitor of the stimulator of interferon genes (STING) used in research to block STING-mediated signaling and inflammatory responses. It inhibits both human and mouse STING by covalently targeting a predicted transmembrane cysteine residue. The compound is supplied as a high-purity solid and as a DMSO solution for cellular assays.
Catalog No. 50-002-59050 Supplier Medchemexpress LLC Supplier No. HY13868310MG
STING-IN-3 is a covalent small-molecule inhibitor of the stimulator of interferon genes (STING) used in research to block STING-mediated signaling and inflammatory responses. It inhibits both human and mouse STING by covalently targeting a predicted transmembrane cysteine residue. The compound is supplied as a high-purity solid and as a DMSO solution for cellular assays.