Learn More
Medchemexpress LLC STING-IN-4 | 2250374-27-5 | C32H46N2O3 | 5 MG
SDP
Catalog No. 5000372476
Encompass_Preferred
STING-IN-4 is a STING inhibitor that inhibits STING expression, thereby reducing the activation of STING and nuclear factor-κB (NF-κB) signaling. It demonstrates anti-inflammatory activity and is suitable for sepsis research.
  • Inhibits STING expression
  • Reduces activation of STING and nuclear factor-κB (NF-κB) signaling
  • Demonstrates anti-inflammatory activity
  • Suitable for sepsis research
  • Inhibits nitric oxide production in RAW264.7 cells
  • Inhibits iNOS expression in RAW264.7 cells
  • Reduces STING degradation
  • May enhance thermal stabilization of STING
  • Inhibits LPS-induced activation of STING/IRF3/NF-κB
  • Protects against LPS-induced liver injuries in mice
  • Inhibits STING/IRF3/NF-κB activation in septic mice livers
Catalog No. 50-003-72476 Supplier Medchemexpress LLC Supplier No. HY1519705MG
STING-IN-4 is a STING inhibitor that inhibits STING expression, thereby reducing the activation of STING and nuclear factor-κB (NF-κB) signaling. It demonstrates anti-inflammatory activity and is suitable for sepsis research.
  • Inhibits STING expression
  • Reduces activation of STING and nuclear factor-κB (NF-κB) signaling
  • Demonstrates anti-inflammatory activity
  • Suitable for sepsis research
  • Inhibits nitric oxide production in RAW264.7 cells
  • Inhibits iNOS expression in RAW264.7 cells
  • Reduces STING degradation
  • May enhance thermal stabilization of STING
  • Inhibits LPS-induced activation of STING/IRF3/NF-κB
  • Protects against LPS-induced liver injuries in mice
  • Inhibits STING/IRF3/NF-κB activation in septic mice livers